missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin - Cy3

Product Code. 10768023
Click to view available options
Quantity:
1 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10768023

Brand: Invitrogen™ 434315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Streptavidin - Cy3

Reactivity: CY3-SA reacts with biotin.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.5 mg/mL
Conjugate Cy3
Quantity 1 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid
Product Line Cy
Product Type Streptavidin Conjugate (fluorescent)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.