missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Pacific Blue™ conjugate

Product Code. 10328392
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10328392

Brand: Invitrogen™ S11222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Invitrogen™'s streptavidin, Pacific Blue™ conjugate is a bright, blue-fluorescent probe with excitation/emission ∼410/455 nm.

Invitrogen™'s streptavidin, Pacific Blue™ conjugate is a bright, blue-fluorescent probe with excitation/emission ∼410/455 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Conjugate Pacific Blue
Description Pacific Blue™ Conjugate Streptavidin, 1mg, Yellow, Solid Form, 407nm Absorbency, Pacific Blue™ Label or Dye, 410/455nm Excitation/Emission, Used in Blot Analysis, Flow Cytometry, Histochemical Applications, Immunoassays, –5 to –30°C Storage Temperature
Detection Method Fluorescence
Formulation Solid
pH 8
Quantity 1 mg
Regulatory Status For research use only (RUO)
Sample Volume 1 mg
Packaging 1 Each
Storage Buffer 10mg/mL PBS
Content And Storage Contains 1 vial of streptavidin Pacific Blue™ conjugate.

Store in freezer (-5 to -30°C) and protect from light.
Excitation/Emission 410/455 nm
Flow Cytometer Laser Lines 405
Shipping Condition Room Temperature
Form Solid
Product Line Pacific Blue
For Use With (Application) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Purity or Quality Grade TLC
Product Type Streptavidin Conjugate
Label or Dye Pacific Blue
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.