Learn More
Novus Biologicals™ Streptavidin Check&Go! Control Kit
Product Code. 18048163 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
30 Tests
Unit Size:
30 tests
Product Code. Quantity unitSize
18048163 30 Tests 30 tests
1 options
This product is for research use only and is not approved for use in humans or in clinical diagnosis
Product Code. 18048163 Supplier Novus Biologicals™ Supplier No. 40040030
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This product is for research use only and is not approved for use in humans or in clinical diagnosis
Control kit
  • Components:
    • 30 Protein A/G strips
    • 2 Cryovials of 10x Running Buffer
    • 1 Vial of 40nm InnovaCoat Gold Biotin
    • 1 Vial of Positive Control
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis
  • Kits are guaranteed for 6 months from date of receipt
TRUSTED_SUSTAINABILITY
Specifications
Kit Contents 30 protein A/G strips, 1 vial of 40 nm InnovaCoat Gold biotin, 1 vial of positive control, 2 cryovials of 10X running buffer
Quantity 30 Tests