missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (APC-Alexa Fluor™ 750)

Product Code. 10678693
Click to view available options
Quantity:
1 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10678693

Brand: Invitrogen™ SA1027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Streptavidin, (APC-Alexa Fluor™ 750)

  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

View a selection guide for all streptavidin RPE, APC, and tandem conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RUO
Color Blue
Concentration 1 mg/mL
Conjugate APC-Alexa Fluor 750
Description APC-Alexa Fluor™ 750 Streptavidin, 1mL, Blue and Clear Solution, APC-Alexa Fluor™ 750 Dye Type, 775nm Emission Wavelength, 600-650nm Excitation Wavelength, PBS Buffer, Fluorescent Detection Method, For Use with Flow Cytometer
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Includes 1 Vial of streptavidin, APC-Alexa Fluor™ 750 conjugate
Quantity 1 mL
Regulatory Status For research use only (RUO)
Packaging Tube
Storage Buffer PBS + BSA
Content And Storage Contains 1 vial of streptavidin, APC-Alexa Fluor™ 750 conjugate.

Store in refrigerator (2°C to 8°C).
Excitation/Emission 600-650/775 nm
Flow Cytometer Laser Lines 633 or 635
Shipping Condition Wet Ice
Form Solution
Product Line Alexa Fluor
For Use With (Application) Cell analysis
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye APC-Alexa Fluor 750
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.