missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin, (APC)
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
Description
- Class: RUO
- Statement: For research use only. Not for use in diagnostic procedures.
- Buffer: PBS
- Preservative: 0.1% Sodium Azide
Specifications
Specifications
| Concentration | 0.1 mg/mL or 0.2 mg/mL |
| Conjugate | APC |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis |
| Quantity | 0.5 mL |
| Content And Storage | Contains 1 vial of streptavidin, APC conjugate. Store in refrigerator (2–8°C). |
| Excitation/Emission | 650/660 |
| Flow Cytometer Laser Lines | 633 or 635 |
| Shipping Condition | Wet Ice |
| Form | Solution |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction