missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin APC Conjugate

Product Code. 15360720
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy
Contains Azide

Product Code. 15360720

Brand: Invitrogen™ 17431782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/mL
Conjugate APC
Quantity 100 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye APC (Allophycocyanin)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.