missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptavidin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£181.00 - £446.00
Specifications
| Antigen | Streptavidin |
|---|---|
| Clone | 1220C |
| Dilution | Western Blot 0.5 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704469
![]() |
R&D Systems
MAB9020-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18705509
![]() |
R&D Systems
MAB9020 |
100 μg |
£446.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Streptavidin Monoclonal specifically detects Streptavidin in Multi-Species samples. It is validated for Western Blot.Specifications
| Streptavidin | |
| Western Blot 0.5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Purified streptavidin from Streptomyces avidinii | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Streptavidin in Western blot. |
| 1220C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Multi-species | |
| SA V1; SA V2; Streptavidin V1; Streptavidin V2 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title