missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptavidin Antibody (1C2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04345AF647
This item is not returnable.
View return policy
Description
Streptavidin Monoclonal antibody specifically detects Streptavidin in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
| Streptavidin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| SA V1; SA V2; Streptavidin V1; Streptavidin V2 | |
| Recombinant Streptavidin (25-183aa) purified from E. coli | |
| 0.1 mL | |
| Biologically Active Proteins, Virology Bacteria and Parasites | |
| Bacteria | |
| Purified |
| Western Blot, ELISA | |
| 1C2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction