missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin, Alexa Fluor™ 660 conjugate
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Alexa Fluor™ 660 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.
Important Features of Alexa Fluor™ 660 Streptavidin Conjugates:
• Alexa Fluor™ 660 streptavidin conjugate has Ex/Em maxima of ∼ (663/690)
• Bright, photostable fluorescence
• High solubility in aqueous solutions
• Available in multiple colors
• Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more
Properties of Alexa Fluor™ Dyes
Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.
Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Related Links:
Learn more about Avidin-Biotin Detection
Learn more about Alexa Fluor™ Dyes
Find out about other Labeled Streptavidin Conjugates
Read Avidin and Streptavidin Conjugates-Section 7.6 in the Molecular Probes Handbook
Important Features of Alexa Fluor™ 660 Streptavidin Conjugates:
• Alexa Fluor™ 660 streptavidin conjugate has Ex/Em maxima of ∼ (663/690)
• Bright, photostable fluorescence
• High solubility in aqueous solutions
• Available in multiple colors
• Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more
Properties of Alexa Fluor™ Dyes
Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.
Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Related Links:
Learn more about Avidin-Biotin Detection
Learn more about Alexa Fluor™ Dyes
Find out about other Labeled Streptavidin Conjugates
Read Avidin and Streptavidin Conjugates-Section 7.6 in the Molecular Probes Handbook
Specifications
Specifications
| Conjugate | Alexa Fluor 660 |
| Quantity | 1 mg |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Shipping Condition | Room Temperature |
| Form | Solid |
| Product Line | Alexa Fluor™ |
| Product Type | Streptavidin Conjugate (fluorescent) |
| Label or Dye | Alexa Fluor Dyes |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction