missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Alexa Fluor™ 633 conjugate

Product Code. 10564813
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10564813 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10564813 Supplier Invitrogen™ Supplier No. S21375
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Streptavidin, Alexa Fluor™ 633 conjugate

Alexa Fluor™ 633 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor™ 633 Streptavidin Conjugates:
• Alexa Fluor™ 633 streptavidin conjugate has Ex/Em maxima of ∼ (632/647)
• Bright, photostable fluorescence
• High solubility in aqueous solutions
• Available in multiple colors
• Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

Properties of Alexa Fluor™ Dyes
Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 633
Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Form Solid
Product Line Alexa Fluor
Product Type Streptavidin Conjugate (fluorescent)
Label or Dye Alexa Fluor Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.