missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin, Alexa Fluor™ 430 conjugate
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Our Alexa Fluor 430 streptavidin conjugate is a bright, green-fluorescent probe (absorption/emission maxima ∼434/539 nm). Streptavidin, a nonglycosylated biotin-binding protein with a near-neutral isoelectric point, reportedly exhibits less nonspecific binding than avidin.
View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
Specifications
Specifications
| Color | Yellow |
| Conjugate | Alexa Fluor 430 |
| Description | Alexa Fluor™ 430 Streptavidin, 1mg, Yellow Solid, Alexa Fluor™ 430 Dye Type, 539nm Emission Wavelength, 434nm Excitation Wavelength, Fluorescent Detection Method, For Use with Blot Analysis, Flow Cytometry, Histochemical Applications, Immunoassays |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Blot analysis, flow cytometry, histochemical applications, immunoassays |
| Formulation | Solid |
| Includes | 1 Vial of streptavidin Alexa Fluor™ 430 conjugate |
| Quantity | 1 mg |
| Regulatory Status | For research use only (RUO) |
| Content And Storage | -5°C to -30°C |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction