missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Alexa Fluor™ 405 conjugate

Product Code. 10412322
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10412322 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10412322 Supplier Invitrogen™ Supplier No. S32351
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Streptavidin, Alexa Fluor™ 405 conjugate

Our streptavidin, Alexa Fluor 405 conjugate is a bright, photostable probe with excitation/emission maxima of 402 and 421 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.

View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 405
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 1 mg
Content And Storage Contains 1 vial of streptavidin Alexa Fluor™ 405 conjugate.

Store in freezer (-5 to -30°C) and protect from light.
Excitation/Emission 402/421
Flow Cytometer Laser Lines 405
Shipping Condition Room Temperature
Form Solid
Product Line Alexa Fluor
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye Alexa Fluor 405
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.