missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Streptavidin
GREENER_CHOICEBIOPHARMA

Product Code. 12873186 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
5 g
Unit Size:
100mg
10mg
1g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12873186 100 mg 100mg
12863186 10 mg 10mg
16491465 1 g 1g
17277730 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12873186 Supplier IBA Lifesciences Supplier No. 20203100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streptavidin is a widely used tool for immobilization and assays of biotinylated proteins or other biotin conjugated molecules.

Streptavidin is a tetrameric protein composed of identic subunits. Each subunit specifically binds one biotin molecule with fM affinity, which makes it one of the strongest non-covalent interactions known. The preparation contains an N- and C-terminal shortened variant (core streptavidin) with improved properties regarding homogeneity, solubility, resistance towards proteolytic degradation, and accessibility of the biotin binding pocket as compared to native streptavidin.

The streptavidin:biotin system is widely used for immobilization and detection of biotinylated molecules, like proteins and nucleic acids. IBA Lifesciences provides bulk amounts of lyophilized streptavidin in reliable and high quality at competitive prices to generate streptavidin-coated surfaces (microplates, SPR chips, beads) or detection reagents conjugated with, e.g., fluorescent dyes.

Please note that streptavidin is not applicable for detection, immobilization, and purification of Strep-tag™II and Twin-Strep-tag™ fusion proteins, since the binding affinity for both tags is too low.

Specifications

Particle Size 13.3 kDa/subunit
Quantity 100 mg
Source Recombinant, expression in E. coli
Purity or Quality Grade >95 %

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.