missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Streptamer™ Solution Set Standard

Product Code. 15260608 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
50 mL washing buffer, 1 mL stock solution
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15260608

Brand: IBA Lifesciences 65603005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Streptamer Solution Set Standard is designed to be used for washing and dissociation.

  • Includes 50 mL Streptamer™ washing buffer (Buffer IS, 10x)
  • Includes 1 mL D-Biotin stock solution for Streptamer™ Technology
  • Both solutions are sterile filtered and do not contain any preservatives
  • Please handle under sterile conditions

Specifications

Includes Washing buffer and D-Biotin stock solution
Quantity 50 mL washing buffer, 1 mL stock solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.