missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences 10x Buffer R - Regeneration Buffer

Product Code. 12638247 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12638247 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12638247 Supplier IBA Lifesciences Supplier No. 21002100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

10x Buffer R is a Strep-Tactin™ regeneration buffer with HABA

  • 10x Buffer R is a 10x concentrated Strep-Tactin™ regeneration buffer containing HABA. HABA (2-[4-hydroxy-benzeneazo]benzoic acid) displaces desthiobiotin after the elution of Strep-tag™II or Twin-Strep-tag™ fusion proteins from the modified biotin-binding pocket of Strep-Tactin™.
  • The yellow buffer turns red upon binding of HABA to Strep-Tactin™, clearly indicating the regeneration process and activity status of the column.
  • Please note that 10x Buffer R is not applicable for regeneration of Strep-Tactin™XT resins, but it can be used to monitor the activity status after the regeneration.

Specifications

Chemical Name or Material Regeneration Buffer
Color Yellow
Concentration 10X
For Use With (Application) For The Regeneration of Immobilized Strep-Tactin
Quantity 100 mL
compliance-icons
Product Identifier
  • Buffer R (10x), Strep-Tactin Regeneration Buffer with HABA
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap and water.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.