Learn More
IBA Lifesciences Strep-Tactin™XT Spin Columns
Product Code. 15712788 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
25 Spin Columns
Unit Size:
Pack of 25
Product Code. 15712788
Brand: IBA Lifesciences 24150025
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
IBA Lifesciences GmbH Strep-Tactin™XT Spin Columns allows for rapid parallel purification of Strep-tag™ and Twin-Strep-tag™ fusion proteins.
  • Strep-Tactin™XT Spin Columns contain dried polymethacrylate, a synthetic material coupled with the streptavidin variant Strep-Tactin™XT and are suitable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • The pre-packed spin columns are provided with receiver tubes and compatible with conventional microcentrifuges, allowing a rapid, parallel processing of multiple samples.
  • Up to 400 μg of mCherry-Strep-tag™II (28 kDa) can be purified under physiological conditions in less than 10 minutes.
  • Prior to application, the spin columns need to be re-hydrated and activated with Buffer W.
  • In contrast to agarose resins, plant cell components such as chlorophyll are not unspecifically bound by the resin, making Strep-Tactin™XT Spin Columns a favorable alternative
  • Please note that the spin columns are not reusable.
Specifications
Quantity 25 Spin Columns