Learn More
IBA Lifesciences Strep-Tactin™XT Coated Microplate
Strep-Tactin™XT coated microplate is a ready-to-use microplate for assay applications
Brand: IBA Lifesciences 2-4101-001
Additional Details : Weight : 0.01100kg
Description
- Ready-to-use Strep-Tactin™XT coated microplate (12 x 8-well strips) for ligand binding assays, immuno assays, enzyme assays, or diagnostic assays under high throughput conditions.
- Due to the high specificity and affinity (nM-pM) of the streptavidin variant Strep-Tactin™, Strep-tag™II or Twin-Strep-tag™ fusion proteins can be selectively and stably captured even from complex mixtures.
- Biomolecules of interest are presented to interacting partners in a uniform manner resulting in reliable and reproducible assays.
- The solid-phase, multi-well format is compatible with standard multichannel pipettes and automated plate readers.
- Twin-Strep-tag™ fusion protein binding capacity determined with bacterial alkaline phosphatase (97 kDa) is 4 pmol for 200 µl incubation volume.
Specifications
1 Plate | |
Microplate |
For Research Use Only.