Learn More
Beskrivning
Due to the high specificity and affinity (nM-pM) of the streptavidin variant Strep-Tactin™ XT, Strep-tag™II or Twin-Strep-tag™, fusion proteins can be selectively and stably captured even from complex mixtures. Biomolecules of interest are presented to interacting partners in a uniform manner resulting in reliable and reproducible assays.
- Ready-to-use Strep-Tactin™XT coated microplate (12 x 8-well strips) for ligand binding assays, immunoassays, enzyme assays, or diagnostic assays under high-throughput conditions.
- The solid-phase, multi-well format is compatible with standard multichannel pipettes and automated plate readers.
- Twin-Strep-tag™ fusion protein binding capacity determined with bacterial alkaline phosphatase (97 kDa) is 4 pmol for 200 μL incubation volume.
Specifikationer
Specifikationer
| Barcode | No Barcode |
| Array | 12 x 8 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Material | Plastic |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Strep-TactinXT |
| Well Shape | Flat Bottom (F-well) |
For research use only.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.