missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ High Capacity Column
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mL
5 mL
10 mL
5 x 0.2 mL
Unit Size:
1 set
Description
- Strep-Tactin™XT 4Flow™ high capacity columns contain a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins.
- The low concentrated agarose enables the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
- In addition, Strep-Tactin™XT 4Flow™ high capacity allows the application of denaturing agents such as urea.
- In comparison to Strep-Tactin™XT 4Flow™, Strep-Tactin™XT 4Flow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 2-fold higher yield of purified protein.
- To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Specifications
| Matrix | 4% agarose, crosslinked |
| Particle Size | 50 to 150 μm |
| Quantity | 5 x 0.2 mL |
| Target | Twin-Strep-Tag™ and Strep-tag™II |
| Format | Pre-packed Gravity Flow Column |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction