missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ Superflow™, 50% Suspension

Product Code. 12866446 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
4 mL
50 mL
Unit Size:
4mL
50mL
This item is not returnable. View return policy

Product Code. 12866446

Brand: IBA Lifesciences 21206025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for purification of recombinant Strep-tag proteins under gravity flow and low pressure or FPLC

  • Strep-Tactin™ Superflow™ is a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.  

Specifications

Quantity 50 mL
Stability 6 months after shipping.
Storage Requirements 2°C to 8°C
Binding Capacity Biotin: >300 nmol/ mL resin
Product Type 50% suspension

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.