Learn More
IBA Lifesciences Strep-Tactin™ Superflow™, 50% Suspension
Product Code. 12886446 Shop All IBA Lifesciences Products
Product Code. 12886446
Brand: IBA Lifesciences 21206500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for purification of recombinant Strep-tag™ proteins under gravity flow and low pressure or FPLC
  • Bulk resin is recommended for packing of columns and cartridges
  • Resin is optimized for column affinity chromatography rather than for batch purification
  • Binding capacity: 1 mL sedimented resin (corresponding to 2 mL of 50% suspension)
  • Useful for one-step purification of 50 to 100 nmol recombinant Strep-tag™ protein (up to 3 mg in case of 30 kDa protein)
Specifications
Product Type Strep-Tactin Superflow