missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ MacroPrep™ Resin

Product Code. 12826516 Shop All IBA Lifesciences Products
Click to view available options
Packaging Quantity:
20 mL
Unit Size:
20mL
This item is not returnable. View return policy

Product Code. 12826516

Brand: IBA Lifesciences 21505010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Strep-Tactin™ MacroPrep™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

  • Strep-Tactin™ MacroPrep™ consists of polymethacrylate, a synthetic material coupled with the streptavidin variant Strep-Tactin™ and is applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • In contrast to agarose resins, plant cell components such as chlorophyll are not unspecifically bound by this resin, which makes Strep-Tactin™ MacroPrep™ the favorable alternative.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary. 

Specifications

For Use With (Application) Purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Form Suspension
Storage Requirements 2°C to 8°C
Binding Capacity Greater than 300 nmol/mL resin (Biotin); 3 mg/mL (Strep-tag™II Binding)
Packaging Quantity 20 mL
Shipping Condition Room temperature

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.