missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17362-1-AP-20UL
This item is not returnable.
View return policy
Description
Involved in spermatogenesis and sperm function. Plays a role in regulation of cell growth. Binds to double-stranded DNA and RNA. Binds most efficiently to poly(I:C) RNA than to poly(dI:dC) DNA. Binds also to single-stranded poly(G) RNA. Binds non-specifically to the mRNA PRM1 3′-UTR and adenovirus VA RNA.Specifications
| STRBP | |
| Polyclonal | |
| Unconjugated | |
| STRBP | |
| ILF3L, p74, SPNR, STRBP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20744, 55342, 84476 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91WM1, Q96SI9, Q9JKU6 | |
| STRBP | |
| STRBP Fusion Protein Ag10720 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction