missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20522
This item is not returnable.
View return policy
Description
STRBP Polyclonal specifically detects STRBP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| STRBP | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| DKFZp434N214, FLJ11307, FLJ14223, FLJ14984, ILF3L, interleukin enhancer binding factor 3-like, MGC21529, MGC3405, p74, spermatid perinuclear RNA binding protein, spermatid perinuclear RNA-binding protein, SPNRFLJ21960 | |
| Rabbit | |
| 74 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (98%), Bovine (95%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Q96SI9 | |
| STRBP | |
| Recombinant protein encompassing a sequence within the N-terminus region of human STRBP. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 55342 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction