Learn More
Phenomenex™ Strata™ C18-E On-Line SPE Column
Product Code. 31745408 Shop All Phenomenex Products
Change view
Click to view available options
Particle Size:
20 μm
50 μm
55 μm
Unit Size:
Each
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
31745408 20 μm Each
31742121 55 μm Each
31743561 50 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31745408 Supplier Phenomenex™ Supplier No. 00MS039B0CB
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Strata SPE products are manufactured to bring you optimal quality, reproducibility, and performance. With over 20 years of experience, Strata SPE products provide clean extracts and high recoveries for a multitude of target analytes across diverse sample matrices.
  • Available retention mechanisms include reversed phase, ion exchange, normal phase and mixed mode
  • 24 phases and counting
  • Cleaner extracts and reproducible results
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 20 μm
Sorbent C18-E
Type Solid Phase Extraction Column