missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRAP Mouse anti-Human, Clone: 5E11F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66712-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in the nucleotide excision repair (NER). This protein was found to be a component of the protein complex that specifically complements the NER defect of xeroderma pigmentosum group C (XP-c) cell extracts in vitro. This protein was also shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, and thus this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.Specifications
| STRAP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y3F4 | |
| STRAP | |
| STRAP Fusion Protein Ag13085 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 5E11F3 | |
| Unconjugated | |
| STRAP | |
| MAP activator with WD repeats, MAWD, PT WD, STRAP, UNR interacting protein, UNRIP, WD 40 repeat protein PT WD | |
| Mouse | |
| Protein G | |
| RUO | |
| 11171 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction