missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ STRAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30067
This item is not returnable.
View return policy
Description
STRAP Polyclonal antibody specifically detects STRAP in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| STRAP | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11171 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| MAP activator with WD repeats, MAWDpt-wd, serine/threonine kinase receptor associated protein, serine-threonine kinase receptor-associated protein, UNR-interacting protein, UNRIPPT-WD, WD-40 repeat protein PT-WD | |
| Between 300 and 350 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction