Learn More
BRAND™ Straight Burette Stopcock with PTFE Key
Product Code. 10461253 Shop All Brand Products
Change view
Click to view available options
Material:
Amber Borosilicate Glass (3.3)
Unit Size:
Each
Product Code. Material Unit Size
10461253 Amber Borosilicate Glass (3.3) Each
1 options
Product Code. 10461253 Supplier BRAND™ Supplier No. 81937
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Straight Burette Stopcock for Burette Cap includes an interchangeable PTFE key.
  • Stopcock housing with precision Borosilicate Glass 3.3 burette tip
  • Interchangeable PTFE key with retention device
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Type Burette Stopcock
Material Amber Borosilicate Glass (3.3)
For Use With (Equipment) 25 mL Burette
Includes PTFE Key