Learn More
Lenz Laborglasinstrumente™ Straight Adapter
Lenz Laborglasinstrumente™ Adapters are manufactured from durable DURAN™ glass tubing.
£10.87 - £15.42
Product Code | Brand | Standard Taper Joint | Joint Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Joint Type | Price | Quantity & Availability | ||||
11846094
|
Lenz Laborglasinstrumente™
05092003 |
NS 24/29 | Cone |
£12.77
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10782921
|
Lenz Laborglasinstrumente™
05092004 |
NS 29/32 | Cone |
£12.83
Each |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11856114
|
Lenz Laborglasinstrumente™
05092002 |
NS 19/26 | Cone |
£11.87
Each |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11866084
|
Lenz Laborglasinstrumente™
05092001 |
NS 14/23 | Cone |
£10.87
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11876184
|
Lenz Laborglasinstrumente™
05101001 |
NS 14/23 | Socket |
£12.14
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11866144
|
Lenz Laborglasinstrumente™
05101004 |
NS 29/32 | Socket |
£15.42
Each |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Straight glass olive
- Available in a range of joint sizes
Specifications
Straight Adapter | |
Duran Glass |