missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRA8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551126
This item is not returnable.
View return policy
Description
STRA8 Polyclonal Antibody detects endogenous levels of STRA8 protein.
Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.
Specifications
| STRA8 | |
| Polyclonal | |
| Unconjugated | |
| STRA8 | |
| RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 346673 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7Z7C7 | |
| STRA8 | |
| Synthesized peptide derived from the Internal region of human Stra8 (aa 151-200). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction