missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRA8 Polyclonal Antibody

Description
The antibody detects endogenous levels of total STRA8 protein.
Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.
Specifications
Specifications
| Antigen | STRA8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | STRA8 |
| Gene Accession No. | Q7Z7C7 |
| Gene Alias | RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8 |
| Gene Symbols | STRA8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?