missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA8 Mouse anti-Human, Rabbit, Mouse, Clone: 1D7E3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | STRA8 |
|---|---|
| Clone | 1D7E3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17669746
|
Proteintech
68071-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17659746
|
Proteintech
68071-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STRA8 Monoclonal antibody apecifically detetects STRA8 antigen in Human, Rabbit, Mouse Samples. It is validated for Western Blot, ELISA.Specifications
| STRA8 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Rabbit, Mouse | |
| NM_182489 | |
| STRA8 | |
| IgG1 | |
| Protein G purification |
| 1D7E3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 346673 | |
| STRA8 fusion protein Ag29680 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title