missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93570-0.02ml
This item is not returnable.
View return policy
Description
STRA13 Polyclonal antibody specifically detects STRA13 in Human samples. It is validated for Western Blot
Specifications
| STRA13 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BHLHE40, CENPX, CENP-X, DEC1, FAAP10, FANCM-interacting histone fold protein 2, Fanconi anemia-associated polypeptide of 10 kDa, HLHB2, MGC14480, MHF2centromere protein X, Retinoic acid-inducible gene D9 protein homolog, stimulated by retinoic acid 13, stimulated by retinoic acid 13 homolog (mouse), Stimulated by retinoic acid gene 13 protein homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-63 of human STRA13 (NP_659435.2). MEGAGAGSGFRKELVSRLLHLHFKDDKTKEAAVRGVRQAQAEDALRVDVDQLEKVLPQLLLDF | |
| 0.02 mL | |
| Direct Reversal of DNA Damage | |
| 201254 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction