missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
STPG4 Polyclonal specifically detects STPG4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | STPG4 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C2orf61 chromosome 2 open reading frame 61 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human STPG4 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?