missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ STP 120 Spin Tissue Processor

Product Code. 11340710 Shop All Epredia Products
Change view
Click to view available options
Includes:
Vent.System, 2 baths, 1 basket
Vent.System, 3 baths, 2 basket
Unit Size:
Each
Product Code. Includes unitSize
11340710 Vent.System, 2 baths, 1 basket Each
12943705 Vent.System, 3 baths, 2 basket Each
2 options
This item is not returnable. View return policy

Product Code. 11340710

Brand: Epredia™ 813150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Process biological specimens from chemical fixation to paraffin infiltration with the Epredia™ STP 120 Spin Tissue Processor.

  • Immediate and delayed start processing modes
  • Programmable spinning speed of either 60 or 70 rpm; spinning can also be programmed off
  • Programmable immersion time in each station (from 1 minute to 99 hours: 59 minutes)
  • Basket capacity of 120 or 240 cassettes
  • Battery backup system in case of power failure

Performance:

  • Reagent vessel tops and charcoal-enhanced ventilation help control processing vapors
  • Up to 10 different processing programs maintained
  • Cassette baskets spin counterclockwise and clockwise within reagent container to improve processing
  • Reagent carryover is reduced through gentle centrifugal spinning of basket above reagent vessel
  • Each reagent vessel holds 1.8 L of processing reagent

Physical Specifications:

  • Electrical Requirements: 100-240 V, 50/60 Hz
  • Diameter: 85 cm (33.5 in.)
  • Height: 50 cm opening to 70 cm (19.6 in. opening to 27.5 in.)
  • Weight: 154 lb. (70 kg)

Includes:

  • Basic instrument
  • Instruction manual
  • Ten reagent vessels and two paraffin baths for single basket operation (120-cassette capacity) or nine reagent vessels and three paraffin baths for double basket operation (240-cassette capacity)
TRUSTED_SUSTAINABILITY

Specifications

Basket Capacity 120 or 240 Cassettes
Reagent Capacity 1.8 L
Includes Vent.System, 2 baths, 1 basket
Diameter (English) 33.5 in.
Diameter (Metric) 85 cm
Height (English) 19.6 to 27.5 in.
Height (Metric) 50 to 70 cm
Weight (English) 154 lb.
Weight (Metric) 70 kg
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.