missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STOX2 Polyclonal Antibody

Product Code. 12809212
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809212 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809212 Supplier Invitrogen™ Supplier No. PA521063

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human kidney tissue lysate. PA5-21063 can be used with blocking peptide PEP-1177.

The Storkhead box protein 2 (STOX2) is the only known paralog to STOX1, a winged-helix domain containing transcription factor believed to play a role in the differentiation of stem cells. STOX2 has been suggested to be part of a molecular profile unique to stem cells, and its mRNA may be part of a transcriptional profile observed with increased inflammatory response to air pollutants. Decreased STOX2 expression levels in decidua are also correlated with preeclampsia, suggesting STOX2 may play a role in the pathophysiology of preeclampsia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STOX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene STOX2
Gene Accession No. Q499E5, Q9P2F5
Gene Alias 4933409N07Rik; AI449080; AI835114; KIAA1392; knockout CG10573-PA related; mKIAA1392; spinelinin; storkhead box 2; storkhead-box protein 2; Stox2
Gene Symbols STOX2
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of human STOX2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306459, 56977, 71069
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.