missing translation for 'onlineSavingsMsg'
Learn More

STOX1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18423026 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423026 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423026 Supplier Novus Biologicals Supplier No. NBP282016FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STOX1 Polyclonal antibody specifically detects STOX1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen STOX1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias C10orf24, PEE4, storkhead box 1, storkhead-box protein 1, winged-helix domain-containing protein
Host Species Rabbit
Immunogen STOX1 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human STOX1. The immunogen is located within amino acids 870 - 920 of STOX1. Amino Acid Squence: SPQNQEMRKHFPQKN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219736
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.