missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
STOX1 Polyclonal antibody specifically detects STOX1 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | STOX1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C10orf24, PEE4, storkhead box 1, storkhead-box protein 1, winged-helix domain-containing protein |
| Host Species | Rabbit |
| Immunogen | STOX1 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human STOX1. The immunogen is located within amino acids 870 - 920 of STOX1. Amino Acid Squence: SPQNQEMRKHFPQKN |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?