Learn More
Specac™ Storm™ 10 Short Pathlength Heated Gas Cell With Controller
Product Code. 15220028
Encompass_Preferred
Change view
Click to view available options
Type:
Storm 10 Heated Gas Cell With Controller
Product Code. Type
15220028 Storm 10 Heated Gas Cell With Controller
1 options
Product Code. 15220028 Supplier Specac™ Supplier No. GS05670
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This Gas Cell enables analysis of gaseous samples and vapours from room temperature to 250°C. The cell is an evacuable stainless steel chamber with an injection septum introduction port and is heated by a low voltage 30V heater surrounding the body. Perfect for gases and vapours at elevated temperatures and vapours generated by solids and liquids at elevated temperatures.
Specifications
Type Storm 10 Heated Gas Cell With Controller
Description This Gas Cell enables analysis of gaseous samples and vapours from room temperature to 250°C.