missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Storm™ 10 Short Pathlength Gas Cell FT-IR NaCl Windows 47mm
Description
Pair of NaCl Windows for use in a 10cm Storm Gas Cell GS05000.
Specifications
Specifications
| For Use With (Equipment) | 10 cm Storm Gas Cell GS05000 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?