Learn More
Specac™ Storm™ 10 Cell Mount for 10cm Gas Cells
Product Code. 15240018
Encompass_Preferred
Change view
Click to view available options
Type:
Storm 10 Cell Mount
Product Code. Type
15240018 Storm 10 Cell Mount
1 options
Product Code. 15240018 Supplier Specac™ Supplier No. GS05030
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
3in. x 2in. Mount Plate to enable a Storm Gas Cell GS05000 to be placed directly into any FTIR instrument.
Specifications
Type Storm 10 Cell Mount
Description 3 x 2 in.