missing translation for 'onlineSavingsMsg'
Learn More

Storkhead Box 2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18419085 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419085 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419085 Supplier Novus Biologicals Supplier No. NBP281978AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Storkhead Box 2 Polyclonal antibody specifically detects Storkhead Box 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Storkhead Box 2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias KIAA1392, Storkhead-Box Protein 2, STOX2, STOX-2
Host Species Rabbit
Immunogen Storkhead Box 2 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human Storkhead Box 2. The immunogen is located within the last 50 amino acids of Storkhead Box 2. Amino Acid Squence: RASAEPPTNEAEKLQKSPN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56977
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.