Learn More
Thermo Scientific™ Store Image and Database Management Software, academic edition
Shop All Thermo Scientific ProductsBeskrivning
Using Thermo Scientific™ Store™ Informatics Software, store and manage data automatically for rapid, highly scalable, global access to all HCS data.
Thermo Scientific™ Store™ software is an SQL Server or Oracle™ database used for storing and managing the data automatically transferred from an HCS instrument. It allows for rapid, highly scalable, global access to all HCS data. Use the Store software after you have collected data and images with the Thermo Scientific™ vHCS Discovery Toolbox.
Highlights:
Highlights:
- Automatic Transfer of Images and Data – Centralized data storage allowing data and image access from multiple locations
- Accessible and Linked – Data is seamlessly linked to its associated image
- Visualize – View images and data linked together and easily compare images
- Export – Built in export formats for software such as Spotfire™, Excel™, and GeneData Screener™ programs
- HCS Connect – Fully open standard, web services API that provides programmatic access to any data and images being managed by Store software
- HCS Gateway – Users can then benefit from a single, open standard repository of images and data regardless of source
Compatible with:
Thermo Scientific™ HCi appliance informatics hardware; Thermo Scientific™ vHCS Discovery Toolbox acquisition and analysis software
Specifikationer
Specifikationer
| Description | Thermo Scientific Store Informatics Software Academic Edition |
| For Use With (Equipment) | ArrayScan High Content Analysis (HCA) Readers, CellInsight CX5 High Content Screening (HCS) Platform |
| Software | Image Management, Database Management |
| License | Academic |
For Research Use Only. Not for use in diagnostic procedures.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?