Learn More
Manutan Storage tray, Gratnells, green,A4 shallow tray
Product Code. 12855506
Encompass_Preferred
Product Code. 12855506 Supplier Manutan Supplier No. A367160
The ever popular Gratnell trays suitable for educational, office and workplace use. Made from extra strong polypropylene, resistant to the majority of chemicals. Flat bottom for easy cleaning. Shallow and jumbo trays have unique cut-out handle with stack and nest facility.
  • A4 shallow type
  • Green
  • Dimension (H x W x D): 75mm x 312mm x 427mm