Learn More
Manutan Storage tray, Gratnells, blue, A3 shallow tray
Product Code. 12854508
Encompass_Preferred
Product Code. 12854508 Supplier Manutan Supplier No. A367145
Only null left
Add to basket
Add to basket
The ever popular Gratnell trays suitable for educational, office and workplace use. Made from extra strong polypropylene, resistant to the majority of chemicals. Flat bottom for easy cleaning. Shallow and jumbo trays have unique cut-out handle with stack and nest facility.
  • A3 shallow type
  • Blue
  • Dimension (H x W x D): 85mm x 453mm x 478mm