missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Storage Solution for Eutech™ EC620130 pH electrode
Description
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes. The Eutech™ Storage Solution is used for the Eutech™ EC620130 pH electrode.
Features:
- Storage solution packed in 500mL durable plastic bottles – economical and convenient for laboratory use.
Specifications
Specifications
| For Use With (Equipment) | EC620130 pH Electrode |
| Type | Storage Solution |
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?