missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz Storage Case

Product Code. 10665195
Click to view available options
:
With 50 Slide Capacity
This item is not returnable. View return policy

Product Code. 10665195

Brand: Heinz Herenz 1060050

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Storage case, black with wooden frame and plastic register. Meant for up to 50 slides.

  • Storage case
  • Black
  • Wooden frame with plastic register
  • For 50 slides

Specifications

Type Microscope Slide Box
Slide Capacity 50
Autoclavable Not Autoclavable
Color Black
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.