missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Storage Box for 80-place Microtube Racks

Product Code. 17597876
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17597876 White Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17597876 Supplier Labcon™ Supplier No. 26012200009
Only null left
Add to basket
Add to basket

This storage box accepts one 80-place rack for freezer storage.

  • Refillable rack with hinged clear lid
  • For use with 80-place tube racks
  • Freezable to -80°C
  • Made in USA

Specifications

Autoclavable Autoclavable
Color White
Positions 80 Positions
Shape Rectangle
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) 80-place Microcentrifuge Tube Racks
Temperature (Metric) Range -80°C to 121°C
Includes Storage Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.