Learn More
Fisherbrand™ Stoppers
Prevents liquid chemical leaks or escape from container.
Brand: Fisherbrand™
Code : 41
Additional Details : Weight : 0.83250kg
Description
- Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
- Preferable for applications that require tighter seal or greater degree of chemical resistance
- Prevent contamination of samples by protecting contents of laboratory glassware from air
- Allows experimenters to shake or mix solutions without spilling them
Specifications
Assorted | |
For laboratory use | |
Rubber |
Stopper | |
67/Pk. |