Learn More
Carl Zeiss™ Stop Slider A, 14 x 40mm
Description
Compatible Microscopes:
- Axio Scope.A1
- Axio Scope.A1 Pol
- Axio Scope.A1 Vario
- Axio Vert.A1 FL
- Axio Vert.A1 MAT
Specifications
Specifications
| Type | Stop Slider A with Aperture/Luminous Field Diaphragm |
| For Use With (Equipment) | Axio Scope; Axio Vert Microscopes |
| Includes | Aperture/luminous field diaphragm |